Eagle™ Floor Model Corrosives Cabinet, Two-Door Model

Provide more space to store hazardous material in your work environment. Eagle Floor Model Corrosives Cabinet can be used for storing containers up to 60 gallons in capacity.

Manufacturer: Eagle™ CRA3010

 View more versions of this product

Catalog No. 19-120-717

  Check Availability
Add to Cart

Alert

Not recommended for use with sulfuric, nitric, or hydrochloric acid.


    Specifications

    Width (English) Interior 40 in.
    Width (Metric) Interior 101.6cm
    Certifications/Compliance FM, NFPA, OSHA
    Capacity (English) 30 gal.
    Depth (English) Interior 15 in.
    Depth (Metric) 45.7cm
    Depth (Metric) Interior 38cm
    Door Style Self Close
    For Use With (Application) Flammables
    Height (English) 44 in.
    Includes HDPE shelves and bottom trays with drip edge
    No. of Doors 2
    No. of Shelves 1
    Surface Coating Epoxy
    Type Storage Cabinet
    Weight (English) 257 lb.
    Weight (Metric) 116.5kg
    Width (English) 43 in.
    Width (Metric) 109cm
    Height (English) Interior 41 in.
    Dimensions (D x W x H) Interior 15 x 40 x 41 in. (38 x 101.6 x 104.1cm)
    Material Steel
    Color Blue
    Depth (English) 18 in.
    Height (Metric) 112cm
    Height (Metric) Interior 104.1cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit