Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eagle™ Floor Model Corrosives Cabinets: Two-Door Models

Provide more space to store hazardous material in your work environment. Eagle Floor Model Corrosives Cabinet can be used for storing containers up to 60 gallons in capacity.

Supplier:  Eagle™ CRA4510X

 View more versions of this product

Catalog No. 19-120-718


Add to Cart

Alert:

Not recommended for use with sulfuric, nitric, or hydrochloric acid.


Description

Description

Provide more space to store hazardous material in your work environment. Eagle Floor Model Corrosives Cabinet can be used for storing containers up to 60 gallons in capacity.

  • Suitable for storage of flammable and nonflammable acids and corrosive liquids
  • Rugged 18-gauge steel cabinets coated with tough blue epoxy powder on interior and exterior surfaces, including shelves, for extra protection against corrosive acids and caustics
  • Coated surfaces are superior to epoxy- or enamel-painted surfaces in case of spill or splash
  • Doors clearly labeled “CAUTION—CORROSIVES”
  • High-density polyethylene trays for shelves and bottom with drip edge contains liquid spills; removable for easy cleaning

Certifications

FM approved.

Compliance

Meet NFPA Code 30 and OSHA requirements.

Specifications

Specifications

45 gal.
Self-close
165.1 cm
157.48 cm
109 cm
2
160.1 kg
Flammables
40 in.
65 in.
FM, NFPA, OSHA
Steel
Blue
Storage Cabinet
2
38 cm
15 x 40 x 62 in. (38 x 101.6 x 157.48 cm)
45.7 cm
101.6 cm
43 in.
62 in.
353 lb.
18 in.
Epoxy
15 in.
HDPE shelves and bottom trays with drip edge
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit