Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eagle™ Floor Model Corrosives Cabinets: Two-Door Models
Provide more space to store hazardous material in your work environment. Eagle Floor Model Corrosives Cabinet can be used for storing containers up to 60 gallons in capacity.
Supplier: Eagle™ CRA47X
Alert:
Not recommended for use with sulfuric, nitric, or hydrochloric acid.
Description
Provide more space to store hazardous material in your work environment. Eagle Floor Model Corrosives Cabinet can be used for storing containers up to 60 gallons in capacity.
- Suitable for storage of flammable and nonflammable acids and corrosive liquids
- Rugged 18-gauge steel cabinets coated with tough blue epoxy powder on interior and exterior surfaces, including shelves, for extra protection against corrosive acids and caustics
- Coated surfaces are superior to epoxy- or enamel-painted surfaces in case of spill or splash
- Doors clearly labeled “CAUTION—CORROSIVES”
- High-density polyethylene trays for shelves and bottom with drip edge contains liquid spills; removable for easy cleaning
Certifications
FM approved.
Compliance
Meet NFPA Code 30 and OSHA requirements.
Specifications
157.48 cm | |
2 | |
165.1 cm | |
45.7 cm | |
2 | |
159 kg | |
Flammables | |
18 in. | |
Blue | |
Manual-close | |
15 in. | |
351 lb. | |
65 in. |
45 gal. | |
Storage Cabinet | |
109 cm | |
101.6 cm | |
15 x 40 x 62 in. (38 x 101.6 x 157.48 cm) | |
38 cm | |
FM, NFPA, OSHA | |
62 in. | |
43 in. | |
Steel | |
40 in. | |
Epoxy | |
HDPE shelves and bottom trays with drip edge |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction