Learn More
Eagle™ Nonmetallic Safety Cans
High-density polyethylene safety cans help reduce danger of explosion from ignition of flammable liquid vapors
Supplier:  Eagle™ 1537
 View more versions of this product
Catalog No. 17-987-404F
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Cans are rust-, corrosion-, and impact-resistant. All metal fittings and flame arresters are stainless steel. Use red cans for flammables; white cans for acids and corrosives.
Type I Safety Cans
  • Feature a single spout with self-closing cap, pressure-relief vent, and flame arrester
  • 2 qt. (2L) and 1 gal. (3.8L) sizes are oval in shape; larger sizes are round
  • Available in red or white
Safety Plunger Cans
  • For dispensing and cleaning of flammable fluids
  • Press lightly on spring-mounted plunger to dispense measured amount of liquid for dampening swab, sponge, or cloth
  • Red only
Certifications
FM approved.
Compliance
Meet OSHA requirements.
Specifications
Specifications
FM, OSHA
Rust and Corrosion Resistant
11.4 L
For use in scientific, industrial and hospital laboratories
Flame arresters, single spout, self-closing cap, pressure-relief vent
32.3 cm
33 mm
2.7 Kg
Type I Safety Can
3 gal.
Red
Polyethylene
12.5 in.
13 in.
6 lb.
Product Suggestions
Product Suggestions