Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eagle™ Nonmetallic Safety Cans

High-density polyethylene safety cans help reduce danger of explosion from ignition of flammable liquid vapors

Manufacturer:  EAGLE MANUFACTURING P714

 View more versions of this product

Catalog No. 17-987-405C


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

1 gal., 3.8L
13 in.
For use in scientific, industrial and hospital laboratories
19.6cm
Rust, corrosion
1.8kg
19.6cm
3.8L
Polyethylene
FM, OSHA
33cm
7.75 in.
Flame arrester
4 lb.
7.75 in.
1 gal.
Red
Plunger Can
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit