Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eagle™ Polyethylene Oily Waste Cans

Designed for temporary storage of solvent, waste rags, and other materials subject to spontaneous combustion

Supplier:  Eagle™ 937FL

 View more versions of this product

Catalog No. 18-009


Add to Cart

Description

Description

Durable, fire-safe cans won't rust or dent and resist most chemical compounds.

  • Cans feature foot treadle that opens cover; self-closing lid
  • Choice of red or high-visibility yellow

Certifications

FM approved.

Compliance

Meet OSHA 29CFR1910.108 and 20CFR1910.106 requirements.

Specifications

Specifications

HDPE
53 L
18 in.
45.7 cm
Red
1 Ea.
7.2 kg
Foot lever
22 in.
Oily Waste Can
FM, OSHA 29 CFR 1910.108, 20 CFR 1910.106
Self Closing
14 gal.
53 cm
16 lb.
55.8 cm
For temporary storage of solvent, waste rags, and other materials subject to spontaneous combustion
21 in.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit