Eagle™ Step-On Biohazard Waste Container Get IT Item

Durable and fire-safe, the Eagle Step-On Biohazard Waste Container is a sound addition to any work environment with hazardous materials.

Manufacturer: Eagle™ 945BIO

 View more versions of this product

Catalog No. 18-880B

  Check Availability
Add to Cart

    Specifications

    Capacity 37.9L
    Certifications/Compliance FM, OSHA 29 CFR 1910.1030, NFPA
    Color Red
    Dimensions (L x W x H) 22 x 18 x 18 in. (56 x 45.7 x 46cm)
    Includes Prominent biohazard warning label, foot lever
    Length (English) 22 in.
    Length (Metric) 56cm
    Material HDPE
    Width (English) 18 in.
    Width (Metric) 45.7cm
    Capacity (English) 10 gal.
    Diameter (English) Outer 18 in.
    Diameter (Metric) Outer 45.7cm
    Height (English) 18 in.
    Height (Metric) 46cm
    Product Type Biohazard Waste Can

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit