Thermo Scientific™ EASY-Column™ Capillary HPLC Column Connector Kits Available on GSA/VA Contract for Federal Government customers only.
Use these connectors, tees and sleeves with Thermo Scientific™ EASY-Column™ Capillary HPLC Columns.
Manufacturer: Thermo Scientific™ SC600
 View more versions of this product
Catalog No. 03-052-617
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Diameter (Metric) 0.79mm
    Description HPLC to column
    For Use With LC/MS
    Type Connector Kit
    Endcapped Yes
    Pore Size 120Å
    Use these connector kits, sleeves, and tees with Thermo Scientific™ EASY-Column™ Capillary Columns. EASY-Columns optimize nanoscale LC and LC/MS with. Easy-to-use, zero dead volume connector kits are available for one- and two-column configurations and ensure simple, reproducible setup for on-line LC/MS. Each column delivers excellent chromatographic performance on any nanoscale LC system. At flow rates of nL/min versus mL/min, the EASY-Column offers higher sensitivity with greater signal-to-noise ratio compared to traditional electrospray.
  • Description & Specifications
Related Products
Product Content Feedback