Learn More
Thermo Scientific™ EASY-Spray™ LC Columns Available on GSA/VA Contract for Federal Government customers only.
Ensure robust nano and capillary flow LC-MS. These LC columns integrate column and emitter and is temperature-controlled for maximum reliability and performance.
Manufacturer:  Thermo Scientific™ ES803
 View more versions of this product
Catalog No. 03-052-574
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Ensure robust nano and capillary flow LC-MS with Thermo Scientific™ EASY-Spray™ LC Columns. The integrated column/emitter design eliminates dead volume and is temperature-controlled for maximum reliability and performance. Rigourously tested to ensure maximum quality, these columns are designed for maximum simplicity and ease-of-use.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
15%
Reversed Phase
Yes
1000bar
2 µm
100 Å
60°C
C18
Analytical Column, EASY-Spray
75 µm
500 mm
Silica, Spherical Fully Porous Ultrapure
2 to 8
450 m²/g
L1
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.