Learn More
XenoTech,LLCSupplier Diversity Partner EASYCYP CYP2C8LR+ 0.5 NMOL DFS Item
Manufacturer:  XenoTech,LLCSupplier Diversity Partner CYP/EZ052
EasyCYP CYP2C8 Low Reductase plus b5 0.5 nmol P450 at 1 nmol/ml, 10 mg/ml protein
Catalog No. 50-142-940