Learn More
STEMCELL Technologies EasySep™ Mouse Pan-DC Enrichment Kit, For processing: 2 x 10^9 cells
SDP
Supplier:  STEMCELL Technologies 19763
Encompass
The EasySep™ Mouse Pan-DC Enrichment Kit is designed to isolate pan-dendritic cells (DCs) from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-pan-DCs and Tetrameric Antibody Complexes recognizing biotin and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.For even faster cell isolations and greater recovery, we recommend the new EasySep™ Mouse Pan-DC Enrichment Kit II (19863) (https://www.stemcell.com/products/easysep-mouse-pan-dc-enrichment-kit-ii.html) which isolates cells in just 13 minutes.
Catalog No. NC1244294
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.