Learn More
Thermo Scientific™ EasyStart™ PCR Mix-in-a-Tube Available on GSA/VA Contract for Federal Government customers only.
Reduce tedious PCR set-up protocols to a single step using Thermo Scientific™ EasyStart™ PCR Mix-in-a-tube.
Manufacturer:  Thermo Scientific™ 6228
 View more versions of this product
Catalog No. 21-402-51
  Check Availability
Add to cart
Specifications
Description
Reduce tedious PCR set-up protocols to a single step using Thermo Scientific™ EasyStart™ PCR Mix-in-a-tube.
Features:
  • Delivers better specificity and yield than conventionally prepared mixtures
  • Compatible with a wide range of enzymes
  • Wax layer provides safe, easy storage of reagent mixture, prevents oxidation and evaporation, and eliminates wasted reagents
Ordering Information:
Use 0.2mL tubes for 0.2mL heating blocks and 0.5mL tubes for 0.5 or 0.6mL heating blocks.
Description & Specifications
Specifications
Strips
15 to 20µL
Tubes can be stored at ambient temperatures for extended periods; To remove PCR product, simply pierce the wax layer with a pipet tip
EasyStart Micro 20 Strips
0.2mL
Thermal Cyclers