Thermo Scientific™ EasyStart™ PCR Mix-in-a-Tube Available on GSA/VA Contract for Federal Government customers only.
Reduce tedious PCR set-up protocols to a single step using Thermo Scientific™ EasyStart™ PCR Mix-in-a-tube.
Manufacturer: Thermo Scientific™ 6028
 View more versions of this product
Catalog No. 21-402-50
  Check Availability
Add to Cart
    Specifications
    Type Tubes
    Volume (Metric) 0.2mL
    Storage Requirements Tubes can be stored at ambient temperatures for extended periods; To remove PCR product, simply pierce the wax layer with a pipet tip
    Description EasyStart Micro 20
    Reaction Volume 15 to 20µL
    Reduce tedious PCR set-up protocols to a single step using Thermo Scientific™ EasyStart™ PCR Mix-in-a-tube.
    Features:
    • Delivers better specificity and yield than conventionally prepared mixtures
    • Compatible with a wide range of enzymes
    • Wax layer provides safe, easy storage of reagent mixture, prevents oxidation and evaporation, and eliminates wasted reagents
    Ordering Information:
    Use 0.2mL tubes for 0.2mL heating blocks and 0.5mL tubes for 0.5 or 0.6mL heating blocks.
  • Description & Specifications
Related Products
Product Content Feedback