Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EB 1089, Tocris Bioscience™

Catalog Number 39931
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
134404-52-7
C30H46O3
454.695
LVLLALCJVJNGQQ-NVSPCIDQSA-N
(1R,3S,5Z)-5-[(2E)-2-[(1R,3aS,7aR)-1-[(1R,2E,4E)-6-Ethyl-6-hydroxy-1-methyl-2,4-octadien-1-yl]-octahydro-7α-methyl-4H-inden-4-ylidene]ethylidene]-4-methylene-1,3-cyclohexanediol
131954623
(1R,3S)-5-[2-[(1R,3aS,7aR)-1-[(2R,3E,5E)-7-ethyl-7-hydroxynona-3,5-dien-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexane-1,3-diol
CCC(CC)(C=CC=CC(C)C1CCC2C1(CCCC2=CC=C3CC(CC(C3=C)O)O)C)O
This item is not returnable. View return policy
134404-52-7
C30H46O3
454.695
LVLLALCJVJNGQQ-NVSPCIDQSA-N
(1R,3S,5Z)-5-[(2E)-2-[(1R,3aS,7aR)-1-[(1R,2E,4E)-6-Ethyl-6-hydroxy-1-methyl-2,4-octadien-1-yl]-octahydro-7α-methyl-4H-inden-4-ylidene]ethylidene]-4-methylene-1,3-cyclohexanediol
131954623
(1R,3S)-5-[2-[(1R,3aS,7aR)-1-[(2R,3E,5E)-7-ethyl-7-hydroxynona-3,5-dien-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexane-1,3-diol
CCC(CC)(C=CC=CC(C)C1CCC2C1(CCCC2=CC=C3CC(CC(C3=C)O)O)C)O

Vitamin D receptor (VDR) agonist

Vitamin D receptor (VDR) agonist; analog of calcitriol (Cat. No. 2551). Exhibits anti-tumor and anti-proliferative activity with reduced hypercalcemic effects. 60 times more potent in inhibiting MCF-7 cell growth in vitro; also shown to induce autophagy in MCF-7 cells. Reverses the effects of parathyroid hormone-related protein (PTHrP).
Quantity 1 mg
Formula Weight 454.68
Percent Purity >98%
Chemical Name or Material EB 1089
Recommended Storage Store at -20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit