Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBAG9/RCAS1 Mouse anti-Human, Clone: CPTC-EBAG9-2, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | EBAG9/RCAS1 |
---|---|
Clone | CPTC-EBAG9-2 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122544
|
Novus Biologicals
NBP30753520UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
EBAG9/RCAS1 Monoclonal specifically detects EBAG9/RCAS1 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
EBAG9/RCAS1 | |
0.2 mg/ml | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Apoptosis | |
10 mM PBS with 0.05% BSA | |
9166 | |
IgG2a κ | |
Protein A or G purified |
CPTC-EBAG9-2 | |
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Cancer-associated surface antigen RCAS1, EB9cancer associated surface antigen, estrogen receptor binding site associated, antigen, 9, Estrogen receptor-binding fragment-associated gene 9 protein, RCAS1PDAF, receptor-binding cancer antigen expressed on SiSo cells | |
Recombinant full-length human EBAG9/RCAS1 protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title