Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ebastine

Click to view available options
Quantity:
1g
5g
90729-43-4
C32H39NO2
469.669
MFCD00865661
MJJALKDDGIKVBE-UHFFFAOYSA-N
ebastine,kestine,ebastel,ebastin,kestin,evastel,bactil,ebastinum latin,las w-090,ebastina spanish
3191
4-(4-benzhydryloxypiperidin-1-yl)-1-(4-tert-butylphenyl)butan-1-one
CC(C)(C)C1=CC=C(C=C1)C(=O)CCCN2CCC(CC2)OC(C3=CC=CC=C3)C4=CC=CC=C4
90729-43-4
C32H39NO2
469.669
MFCD00865661
MJJALKDDGIKVBE-UHFFFAOYSA-N
ebastine,kestine,ebastel,ebastin,kestin,evastel,bactil,ebastinum latin,las w-090,ebastina spanish
3191
4-(4-benzhydryloxypiperidin-1-yl)-1-(4-tert-butylphenyl)butan-1-one
CC(C)(C)C1=CC=C(C=C1)C(=O)CCCN2CCC(CC2)OC(C3=CC=CC=C3)C4=CC=CC=C4

A histamine H1 receptor antagonist

Solubility
Slightly soluble in chloroform, methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Color White
Melting Point 84°C to 87°C
Quantity 5g
Merck Index 14,3484
Formula Weight 469.66
Physical Form Powder
Chemical Name or Material Ebastine

RTECSNumber : EL8140000