Learn More
Eberbach Blending Assembly, Semi Micro
For Waring™ blenders
Manufacturer:  Eberbach E8730.00
Catalog No. 14-509-33B
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • 32psi held for extended period
  • Stainless steel blade; Other part corrosion resistant chromium plated brass made
  • Nitrile seal
  • Dia.: 2.36 cm (0.93 in.)
Specifications
Specifications
Blending Assembly
Semi micro
Waring™ and Eberbach blending container
Stainless Steel