Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBF-3 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$436.00

Specifications

Antigen EBF-3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP182378
View Documents
Novus Biologicals
NBP182378
100 μL
Each of 1 for $436.00
Add to cart
 
Description

Description

EBF-3 Polyclonal specifically detects EBF-3 in Rat samples. It is validated for Western Blot.
Specifications

Specifications

EBF-3
Polyclonal
Rabbit
NP_001101976
253738
Synthetic peptide towards Ebf3. Peptide sequence MNPVRSWMHTAGVVDANTAAQSGVGLARAHFEKQPPSNLRKSNFFHFVLA.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
RUO
COE3OE-2, DKFZp667B0210, early B-cell factor 3O/E-2, EBF-3, Olf-1/EBF-like 2, transcription factor COE3
Ebf3
IgG
Affinity Purified
65 kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit