Learn More
EBI3 Mouse anti-Human, Alexa Fluor™ 532, Clone: 15k8D10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP227362AF532
Catalog No. NP227362A32
Add to cart
Description
Description
EBI3 Monoclonal antibody specifically detects EBI3 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
EBI3
15k8D10
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
cytokine receptor, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 subunit beta, IL27B, IL-27B, interleukin-27 subunit beta
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
The human Interleukin-35 protein is a dimeric protein consisting of the human IL-12a/p35 subunit (NP_000873.2) and the human EBI3 (NP_005746) protein. This clone has been tested to be specific to the EBI3 subunit of the target protein. Clone 15K8D10 tests as being specific to the EBI3 subunit.
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
EBI3
A full-length human Interleukin-35-Fc fusion protein was used as immunogen for this antibody.
0.1 mL
Primary
10148
Human