Learn More
EBI3 Rat anti-Human, Mouse, Clone: DNT27, Abnova™
Rat monoclonal antibody raised against mouse Ebi3.
Manufacturer:  Abnova Corporation MAB12645
Catalog No. 89-934-057
Add to cart
Description
Description
Specifications
Specifications
EBI3
DNT27
Rat monoclonal antibody raised against mouse Ebi3.
In aqueous buffer (Stabilizer, 0.09% sodium azide).
EBI-3/IL-27
Rat
IgG2a, kappa
50 ug
Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Murine
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
Ebi3
Mouse Ebi3
Affinity purification
RUO
Primary
50498