Learn More
Invitrogen™ eBioscience™ Phytohemagglutinin-L (PHA-L) Solution (500X)
Click to view available options
Quantity:
100 μL
4 x 100 μL
Description
- A ready to use solution of PHA-L in aqueous buffer
- PHA is a mixture of lectins isolated from the red kidney bean, Phaseolus vulgaris
- It is the fraction of PHA that exhibits strong leukoagglutinating and mitogenic activity, but very little erythroagglutinating activity
- This reagent is intended for use in in vitro activation of human and mouse leukocytes
Specifications
Specifications
Quantity | 4 x 100 μL |
Product Line | eBioscience™ |
Product Type | Phytohemagglutinin-L (PHA-L) Solution |
For Research Use Only. Not for use in diagnostic procedures.