Learn More
EBLN2 Mouse anti-Human, Clone: OTI2D5, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808871
Catalog No. 50-166-9139
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
May act as an RNA-binding protein. The C-terminal region is highly homologous to the bornavirus nucleocapsid N protein that binds viral RNA and oligomerizes. The viral protein also possesses a nuclear import and a nuclear export signal. These 2 signals seem absent in EBLN-2 supporting an unrelated function in Human.
Specifications
Specifications
EBLN2
Monoclonal
Unconjugated
EBLN2
Lyophilized
EBLN2
Full length human recombit protein of human EBLN2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI2D5
PBS with 8% trehalose and no preservative; pH 7.3
Q6P2I7
EBLN-2
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
55096