Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ebola Virus VP40 (subtype Bundibugyo, strain Uganda 2007) Monoclonal Antibody (04)
GREENER_CHOICE

Catalog No. PIMA529996
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA529996 100 μL
1 options

Catalog No. PIMA529996

Supplier: Invitrogen™ MA529996

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) VP40/Matrix protein VP40.

Ebola virus is an enveloped, negative-stranded RNA virus. This virus belongs to the family Filoviridae and has a distinctive morphology similar only to that of Marburg virus, but apparently has no antigenic characteristics of Marburg virus. Its genome is a nonsegmented RNA strand. SDS-PAGE profiles demonstrate the presence of seven structural proteins. These are the major nucleoprotein NP (104 K), glycoprotein GP (125K), L protein (180 K), and four proteins: VP40 (40 K), ViPX (35 K), VP30 (30 K) and VP24 (24 K). All the proteins are encoded by monocistronic mRNA transcripts complementary to virion RNA. GP is the major protein of the surface spikes and only this 125K viral protein is glycosylated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ebola Virus VP40 (subtype Bundibugyo, strain Uganda 2007)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 04
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias EVD
Host Species Mouse
Immunogen Recombinant Ebola virus EBOV (subtype Bundibugyo, strain Uganda 2007) VP40/Matrix protein VP40 Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.