Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EBOV VP30 protein Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313345
This item is not returnable.
View return policy
Description
EBOV VP30 protein Polyclonal antibody specifically detects EBOV VP30 protein in Virus samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| EBOV VP30 protein | |
| Polyclonal | |
| PBS, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Capsid protein VP30, EbolaVP30, eVP30, Minor nucleoprotein VP30, Transcriptional activator VP30, Viral protein 30 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Zaire ebola virus VP30 (Zaire ebolavirus (strain Mayinga-76)). The exact sequence is proprietary. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction