Learn More
Trajan Scientific and Medical™ EC Science Silicon Septa
Designed for long lifetime and low injection port adhesion.
Supplier: Trajan Scientific and Medical™ 041905
Description
- Combines long injection lifetime, low bleed and low injection port adhesion
- Reliable seal against carrier gas pressure in the inlet
- No bleed or contamination into the GC system
- Maximum operating temperature of 350°C (300°C for 17mm size)
Specifications
EC | |
Plug | |
50/Pk. |