Learn More
Trajan Scientific and Medical™ EC Science Silicon Septa
Designed for long lifetime and low injection port adhesion.
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
04-190-5
|
Trajan Scientific and Medical™
041905 |
Plug | Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | View substitute products » | |||||
Description
- Combines long injection lifetime, low bleed and low injection port adhesion
- Reliable seal against carrier gas pressure in the inlet
- No bleed or contamination into the GC system
- Maximum operating temperature of 350°C (300°C for 17mm size)
Specifications
EC |