Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECAT1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548144

Catalog No. PA548144


Add to cart

Description

Description

The protein encoded by this gene belongs to the KHDC1 family, members of which contain an atypical KH domain that may not bind RNA like canonical KH domains. This gene is specifically expressed in the oocytes, and recent studies suggest that it may function as a regulator of genomic imprinting in the oocyte. Mutations in this gene are associated with recurrent biparental complete hydatidiform mole. [provided by RefSeq, Dec 2011]
Specifications

Specifications

ECAT1
Polyclonal
PBS with 0.09% sodium azide
Q587J8
KHDC3-like protein, ES cell-associated transcript 1 protein, KHDC3L, C6orf221, ECAT1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
154288
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
KHDC3L
Liquid
KHDC3L
KLH conjugated synthetic peptide between 20-48 amino acids from the N-terminal region of human ECAT1.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit