Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECE-1 Antibody (OTI2B6), DyLight 350, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270612UV

Catalog No. NBP270612UV


Add to Cart

Description

Description

ECE-1 Monoclonal specifically detects ECE-1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

ECE-1
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
ECE1
Recombinant protein fragment corresponding to amino acids 486-770 of human ECE-1 (NP_001388) produced in E.coli.
0.1 mL
1889.0
Store at 4°C in the dark.
Western Blot
OTI2B6
Western Blot
EC 3.4.24, EC 3.4.24.71, ECE, ECE-1, endothelin converting enzyme 1, endothelin-converting enzyme 1
Mouse
Affinity Purified
Primary
Human, Mouse, Rat
IgG1
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit