Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECE-1 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI2B6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270612AF405

Catalog No. N270612A405


Add to cart

Description

Description

ECE-1 Monoclonal antibody specifically detects ECE-1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ECE-1
OTI2B6
Western Blot
Affinity Purified, Conjugated
ECE1
Recombinant protein fragment corresponding to amino acids 486-770 of human ECE-1 (NP_001388) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
EC 3.4.24, EC 3.4.24.71, ECE, ECE-1, endothelin converting enzyme 1, endothelin-converting enzyme 1
Mouse
IgG1
0.1 mL
Primary
1889.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit