Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ECE2 Monoclonal Antibody (233914)

Rat Monoclonal Antibody

Supplier:  Invitrogen™ MA523941

Catalog No. PIMA523941


Only null left
Add to Cart

Description

Description

In Western blots, 30% cross-reactivity with recombinant human (rh) ECE-1 and no cross-reactivity with rhKELL is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Converts big endothelin-1 to endothelin-1. Also involved in the processing of various neuroendocrine peptides, including neurotensin, angiotensin I, substance P, proenkephalin-derived peptides, and prodynorphin-derived peptides. May limit beta-amyloid peptide accumulation in brain. May also have methyltransferase activity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ECE2
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P0DPD6
Ece2
Mouse myeloma cell line NS0-derived recombinant human ECE-2 Val104-Leu787.
100 μg
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
233914
Unconjugated
Ece2
1810009K13Rik; 6330509A19Rik; 9630025D12Rik; BB127715; Ece2; ECE-2; ECE-2a-1; ECE-2a-2; ECE-2b-1; ECE-2b-2; EEF1A lysine methyltransferase 4; EEF1AKMT4; EEF1AKMT4-ECE2; EEF1AKMT4-ECE2 readthrough transcript protein; endothelin converting enzyme 2; endothelin converting enzyme-2; endothelin-converting enzyme 2; Endothelin-converting enzyme 2 region; endothelin-converting enzyme 2a-1; endothelin-converting enzyme 2a-2; endothelin-converting enzyme 2b-1; endothelin-converting enzyme 2b-2; KIAA0604; Methyltransferase-like region; UNQ403/PRO740
Rat
Protein A/G
RUO
9718
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.