Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ECE2 Polyclonal Antibody
ECE2 Polyclonal Antibody
Supplier: Thermo Scientific PA566470
Description
ECE2 Polyclonal Antibody for ICC/IF

Specifications
ECE2 | |
Polyclonal | |
Unconjugated | |
Ece2 | |
1810009K13Rik; 6330509A19Rik; 9630025D12Rik; BB127715; Ece2; ECE-2; ECE-2a-1; ECE-2a-2; ECE-2b-1; ECE-2b-2; EEF1A lysine methyltransferase 4; EEF1AKMT4; EEF1AKMT4-ECE2; EEF1AKMT4-ECE2 readthrough transcript protein; endothelin converting enzyme 2; endothelin converting enzyme-2; endothelin-converting enzyme 2; Endothelin-converting enzyme 2 region; endothelin-converting enzyme 2a-1; endothelin-converting enzyme 2a-2; endothelin-converting enzyme 2b-1; endothelin-converting enzyme 2b-2; KIAA0604; Methyltransferase-like region; UNQ403/PRO740 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9718 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P0DPD6 | |
Ece2 | |
Recombinant Human ECE2. Recombinant protein control fragment (Product #RP-107145). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction