Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ECFP-Tag Monoclonal Antibody (10H5), HRP, Bioss

Mouse Monoclonal Antibody
Supplier: Bioss BSM33189MHRP

This item is not returnable.
View return policy
Description
ECFP-Tag Monoclonal antibody specifically detects ECFP-Tag in Tag samples. It is validated for Western BlotSpecifications
ECFP-Tag | |
Monoclonal | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
Mouse | |
Protein G | |
RUO | |
-20°C or -80°C if preferred | |
Liquid | |
Tag |
Western Blot | |
10H5 | |
HRP | |
eCFP | |
Recombinant ECFP | |
100 μL | |
Primary | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction