Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECH1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15473820UL

 View more versions of this product

Catalog No. NBP15473820


Add to cart

Description

Description

ECH1 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ECH1
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
ECH1
Synthetic peptides corresponding to ECH1(enoyl Coenzyme A hydratase 1, peroxisomal) The peptide sequence was selected from the N terminal of ECH1. Peptide sequence PDHSYESLRVTSAQKHVLHVQLNRPNKRNAMNKVFWREMVECFNKISRDA.
36 kDa
20ul
Lipid and Metabolism
Primary
1891
Western Blot
Western Blot 1:100-1:2000
Q13011
delta(3,5)-Delta(2,4)-dienoyl-CoA isomerase, mitochondrial, delta3,5-delta2,4-dienoyl-CoA isomerase, dienoyl-CoA isomerase, EC 5.3.3.-, enoyl CoA hydratase 1, peroxisomal, enoyl Coenzyme A hydratase 1, peroxisomal, HPXEL, peroxisomal enoyl-CoA hydratase 1
Rabbit
IgG
Protein A purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit