Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECHDC2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP179298

 View more versions of this product

Catalog No. NBP179298


Add to cart

Description

Description

ECHDC2 Polyclonal antibody specifically detects ECHDC2 in Human, Mouse, Rat, Porcine, Bovine, Equine, Guinea Pig, Rabbit, Zebrafish samples. It is validated for Western Blot.
Specifications

Specifications

ECHDC2
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
ECHDC2
Synthetic peptide directed towards the middle region of human ECHDC2The immunogen for this antibody is ECHDC2. Peptide sequence TQRLPRCLGVALAKELIFTGRRLSGTEAHVLGLVNHAVAQNEEGDAAYQR.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Expected identity based on immunogen sequence: Human: 100%; Rabbit: 100%; Rat: 100%; Mouse: 92%; Chicken: 85%; Guinea pig: 85%.
Bovine, Equine, Guinea Pig, Human, Mouse, Porcine, Rabbit, Rat, Zebrafish
Western Blot
Western Blot 1:1000
NP_060751
DKFZp686F0985, DKFZp686K13244, enoyl CoA hydratase domain containing 2, enoyl Coenzyme A hydratase domain containing 2, FLJ10948, FLJ45240, FLJ52213, FLJ52450, FLJ78805, FLJ99576, MGC57155, mitochondrial
Rabbit
IgG
100 ul
Lipid and Metabolism
Primary
55268
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit