Learn More
Torrey Pines Scientific™ EchoTherm™ Benchtop Chilling/Heating Incubator Sample Blocks
Manufacturer:  Torrey Pines Scientific™ 6205012
 View more versions of this product
Catalog No. 11-680-24
Add to cart
Description
Description
Blocks are machined from solid aluminum to fit precisely the tube size and shape. Block bottoms are also machined to nest on the incubator plate to prevent movement. Feature an anodized black finish for better heat transfer.
Specifications
Specifications
Echotherm benchtop chilling/heating incubator
Aluminum
For 0.5mL centrifuge tube
Block