EchoTherm™ Benchtop Incubator, Chiller and Freezer
Five devices in one—water bath, dry bath, incubator, refrigerator, and freezer; Heats, chills, or freezes samples from -10° to +100°C—even at ambient temperatures; Energy-efficient Peltier design—no compressor or CFCs.
$1687.41
Includes: Includes RS-232-C interface for computer-directed data collection and unit control.A built-in datalogger can be programmed to collect temperature data points in 1-second, 1-minute, or 5-minute intervals to a total of 8110 data points. Includes a universal power supply that will accept inputs from 85 to 265 VAC, 50/60Hz and 12VDC output to chiller/heater module.
  • Products 1
  • Description & Specifications
Products
Catalog Number Mfr. No. Description Price Quantity Availability  
11-680-22
Required Accessories
Torrey Pines Scientific
IC20
Incubator-Chiller-Freezer
Each for $1,687.41
Check Availability
Add to Cart
Descriptions & Specifications
Related Products