Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Torrey Pines Scientific™ EchoTherm™ Chilling/Heating Dry Baths

Catalog No. p-2330837
Click to view available options
Model:
IC22
IC25
Voltage:
100/260 V
115 V
230 V
Wattage:
50 w
100 w
Weight (English):
8 lb.
13 lb.

Includes

Units come complete with bench-top power supply, instruction manual and AC line cord. Sample blocks and covers are available as accessories and are shown on the price list.

Chill, heat or freeze two separate sets of samples at two separate temperatures with individual control from -10° to 100°C

The IC22 2-Position Electronic Chilling/Heating Dry Bath is versatile, reliable and easy to use. Each heating/chilling station has its own Peltier module, and the entire unit has only two moving parts: the fans. Each plate can be set separately for any temperature within the range of unit without interaction between the plates. The IC22 has simple digital controls and a two-line alphanumeric digital display that is backlit for easy viewing. The display shows the target and actual temperatures of both plates.

 

  • Energy-efficient Peltier design
  • Digital control to 1°C from -20 to 100°C
  • Two separate temperatures on separate plates
  • Sample blocks for 0.2, 0.5 and 1.5mL centrifuge tubes, 96-well and 384-well assay plates, 2mL vials, various sizes of test tubes and more
  • Control at room temperature
  • Two countdown timers (up to 30 days each) with alarm
  • RS232 I/O port for control or data collection
  • Two data loggers
  • Electronic calibration
  • Compact size; small footprint on the bench
Specifications
  • Temperature Range on each plate: -20° to 100°C
  • Readability: 1°C
  • Accuracy: ±1°C
  • Stability: ±1°C
  • Sensor: Platinum RTD
  • Controller Type: PID
  • Uniformity: ±0.5°C

 

 

  • Maintaining 14°C for ligation reactions
  • Maintaining 17°C for storing oocytes
  • Storing samples at ice bucket temperatures
  • Incubate samples in 96-well assay plates, or in aluminum blocks holding 0.2, 0.5 or 1.5mL centrifuge tubes
  • Cooling blood samples prior to coagulation testing
  • Storing enzymes or DNA libraries at your work station
  • Transporting samples from the field at exactly the right temperature
  • Chilling or heating samples in the field
  • Enzyme reactions and deactivations
  • Freezing and storing most any small sample at your workstation
  • PCR sample preparation
  • Hybridizations
  • General molecular biology applications

 

Compliance

UL, CSA and CE compliant

Warranty and Services

One year

Specifications

Certifications/Compliance UL, CSA, CE
Material Aluminum
Temperature Range (Metric) -20°C to +100°C
Type Dry Bath Block
Display Digital
Includes Bench top power supply, instruction manual and AC line cord, sample blocks and covers
Model IC22
Run Time 00:01 sec. to 30 days
Temperature Accuracy ±1°C
Temperature Uniformity ±0.5°C
Warranty 1 Year
Width (English) 6.5 in.
Width (Metric) 16.5 cm
Height (English) 3.5 in.
Height (Metric) 8.9 cm
Weight (English) 13 lb.
Weight (Metric) 5.85 kg
Electrical Requirements 115 V, 50/60 Hz
Voltage 115 V
Wattage 100 w
Amperage 2 A
Frequency 50/60 Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.