ECL Plex Goat anti-Mouse IgG-Cy™ 5 Secondary Antibody, GE Healthcare

Manufacturer: GE Healthcare Life Sciences PA45010

 View more versions of this product

Catalog No. 45-001-211

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Antigen IgG
    Format Purified
    Host Species Goat
    Isotype IgG
    Quantity 600μg
    Primary or Secondary Secondary
    Monoclonal or Polyclonal Monoclonal
    Conjugate Cy5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit