Learn More
BioVision IncSupplier Diversity Partner ECL Western Blotting Substrate Kit: Superior Signal, clear background DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K82050
BioVision ECL Western Blotting Substrate is a highly sensitive, nonradioactive, enhanced luminol-based chemiluminescent substrate for easy detection of horseradish peroxidase (HRP) on immunoblots. BioVision ECL Western Blotting Substrate offers superior signal and clear background. Blots can be repeatedly exposed to X-ray film or a CCD camera to obtain optimal results or stripped of the immunodetection reagents and re-probed. The special formulation of BioVision ECL substrate makes it ideal substitute for Amersham ECL substrate without the need for additional optimization of assay conditions. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9935