Learn More
BioVision IncSupplier Diversity Partner SuperBrite™ ECL Western Blot Substrate/Detection Kit: Highly Sensitive, Nonradioactive, Enhanced luminol-based chemiluminescent Substrate for western and dot blots. Superior signal and Clear background. Pack Size: 50 ml. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K82450
BioVision’s SuperBrite™ ECL kit includes second-generation ECL substrates. They are highly sensitive, non-radioactive, and enhanced luminol-based chemiluminescent substrates for easy detection of horseradish peroxidase (HRP) on immunoblots. Blots can be repeatedly exposed to X-ray film or a CCD camera to obtain optimal results or stripped off the immune-detection reagents and re-probed. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9939