Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECM-1/Secretory Component P85 Mouse anti-Human, Rat, Clone: rSPM217, Novus Biologicals™

Mouse Monoclonal Antibody

$159.82 - $374.34

Specifications

Antigen ECM-1/Secretory Component P85
Clone rSPM217
Concentration 0.2 mg/ml
Dilution Flow Cytometry 0.5-1 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122038
View Documents
Novus Biologicals
NBP30728220UG
20 μg
Each for $159.82
Add to cart
 
NB122037
View Documents
Novus Biologicals
NBP30728210UG
100 μg
Each for $374.34
Add to cart
 
Description

Description

ECM-1/Secretory Component P85 Monoclonal antibody specifically detects ECM-1/Secretory Component P85 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

ECM-1/Secretory Component P85
0.2 mg/ml
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Purified
RUO
Human, Rat
ECM1, extracellular matrix protein 1, Secretory Component Glycoprotein, Secretory component p85
Recombinant full-length human ECM-1/Secretory Component P85 protein
Monoclonal
Protein A or G purified
rSPM217
Flow Cytometry 0.5-1 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin
Unconjugated
Mouse
Extracellular Matrix, Immunology
10 mM PBS with 0.05% BSA
1893
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit