Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ECM2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557239
Description
Immunogen sequence: EGEEDEEDEE DPVRGDMFRM PSRSPLPAPP RGTLRLPSGC SLSYRTISCI NAMLTQIPPL TAPQITSLEL TGNSIASIPD EAFNGL Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 69%.
ECM2 encodes extracellular matrix protein 2, so named because it shares extensive similarity with known extracelluar matrix proteins. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
ECM2 | |
Polyclonal | |
Unconjugated | |
ECM2 | |
9030618O22Rik; BC065151; ECM2; Extracellular matrix protein 2; extracellular matrix protein 2, female organ and adipocyte specific; extracellular matrix protein 2-like; LOC100361383; Matrix glycoprotein SC1/ECM2; Tenonectin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1842 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94769 | |
ECM2 | |
Recombinant protein corresponding to Human ECM2. Recombinant protein control fragment (Product #RP-92900). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction