Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ eco (Escherichia coli) Recombinant Protein

Used for SDS-PAGE

Manufacturer:  Abnova™ P4873

Catalog No. 89-942-730


Add to cart

Description

Description

Sequence: MGSSHHHHHHSSGLVPRGSHMAESVQPLEKIAPYPQAEKGMKRQVIQLTPQEDESTLKVELLIGQTLEVDCNLHRLGGKLENKTLEGWGYDYYVFDKVSSPVSTMMACPDGKKEKKFVTAYLGDAGMLRYNSKLPIVVYTPDNVDVKYRVWKAEEKIDNAVVR
Specifications

Specifications

NP_416713
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
18.3kDa
Escherichia coli expression system
>95% by SDS-PAGE
50 ug
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
ECK2201|JW2197|eti
eco
Escherichia coli
1 mg/mL
Liquid
946700
eco (Escherichia coli) Recombinant protein
Conventional Chromatography
15% SDS-PAGE Stained with Coomassie Blue
MGSSHHHHHHSSGLVPRGSHMAESVQPLEKIAPYPQAEKGMKRQVIQLTPQEDESTLKVELLIGQTLEVDCNLHRLGGKLENKTLEGWGYDYYVFDKVSSPVSTMMACPDGKKEKKFVTAYLGDAGMLRYNSKLPIVVYTPDNVDVKYRVWKAEEKIDNAVVR
RUO
eco
Escherichia coli expression system
His
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit