Learn More
Brady™ Eco Friendly Paperboard Danger Sign: FLAMMABLE LIQUIDS
Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.
Specifications
Color | Red on White |
---|---|
Shape | Rectangular |
Displayed Text | FLAMMABLE LIQUIDS |
Temperature Range (English) | -40°F to 175°F |
Finish | Not Laminated |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19-104-853
|
Brady™
115955 |
10 x 7 in. | Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | |||||
19-104-852
|
Brady™
115954 |
14 x 10 in. | Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | |||||
Description
- Paperboard material is abrasion-resistant and chemical-resistant
- Bold and bright, providing high visibility to critical messages
- For indoor use
Specifications
Red on White | |
FLAMMABLE LIQUIDS | |
Not Laminated | |
1 Ea. | |
Paper board | |
175°F | |
0.14 in. |
Rectangular | |
-40°F to 175°F | |
Paper | |
Non-adhesive | |
Corner holes | |
-40°F |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.