Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZING™ Enterprises Eco GHS Projecting Sign, SDS Station

ZING Enterprises Eco Safety sign available in V-Style, L-Style, Standard TriView, and Slim Triview signage.

Manufacturer:  ZING™ Enterprises 6054

 View more versions of this product

Catalog No. 18-001-879


Add to cart

Description

Description

Features:

  • Identifies designated SDS information areas
  • Supports compliance with the new GHS/Hazard Communication requirements.
  • Significantly increases visibility of information
  • Made from high quality 70% post-consumer recycled plastic
  • UL Validated for recycled content which substantiates material compliance with various green codes, standards, and procurement policies including the LEED Green Building Rating System, IgCC, GSA Advantage, ASHRAE 189.1 and others.
  • Recycled material supports sustainability initiatives and LEED credits
  • Includes 2 mounting holes and mounting adhesive tabs and measures.

Certifications

GHS OSHA Hazard Communication 2012

Specifications

Specifications

Red on White
Plastic
7in x 2.5in
Communication
SDS Station
English
1
Corner Holes
7in.
2.5in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit