Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZING™ Enterprises Eco GHS-SDS Portable Document Case, Recycled Stainless Steel

SDS centers support compliance with GHS/Hazard Communication requirements.

Manufacturer:  ZING™ Enterprises 6046

Catalog No. 18-002-964


Add to cart

Description

Description

ZING Eco SDS Portable Document Case stores important safety and training documents to comply with the new GHS/Hazard Communication requirements. Case is highly visible with bright red/white colors and includes a lockable hasp on the door for extra security.  Document case is designed with a built-in 3-ring holder that stores up to 300 sheets and can be conveniently carried to the job site or mounted to a wall. Case is constructed from heavy gauge 75% recycled stainless steel that reinforces sustainability initiatives as well as compliance. Recycled materials also support LEED credits. Measures 12 in.H x 13-5/8 in.W x 3 in.D. Made in the USA.

Specifications

Specifications

1
Safety
13.5 in.
Stainless Steel
13.5 in. x 3.5 in.
Red
Hazard Communication
Floor standing
3.5 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit