Learn More
ZING™ Enterprises Eco Parking Sign Handicapped Parking Fine Missouri
ZING Enterprises Eco Safety sign complies with state laws to designate access to handicap parking spaces.
Manufacturer:  ZING™ Enterprises 2693
 View more versions of this product
Catalog No. 18-002-093
Add to cart
Description
Description
Features:
  • Highly visible with 3M Engineer Grade Prismatic Reflective Sheeting
  • Meets MUTCD Standards
  • Made from high quality 99% recycled .080 Aluminum (80% Post Consumer) that is UL Validated for recycled content which substantiates material compliance with various green codes, standards, and procurement policies including the LEED Green Building Rating System, IgCC, GSA Advantage, ASHRAE 189.1 and others.
  • Recovered materials also exceed EPA's recommended content levels for signage and are EPA CPG Listed
  • Recycled materials support sustainability initiatives and LEED Certified Building Projects
  • Includes pre-drilled holes for easy mounting
  • Made in the USA.
Certifications
State ADA Compliance, Meets MUTCD Standards
Specifications
Specifications
White, Blue
Engineer Grade Prismatic
6in x 12in
Communication
$50 To $300 Fine
English
1
Corner Holes
6in
12in