Learn More
ZING™ Enterprises Eco Parking Sign Handicapped Parking Permit
ZING Enterprises Eco Safety Sign is pre-drilled for easy mounting. Available in two grades of reflective sheeting.
Manufacturer:  ZING™ Enterprises 2212
 View more versions of this product
Catalog No. 18-002-063
Add to cart
Description
Description
3M Engineer Grade Prismatic sheeting
  • Recommended for commercial signs and non-critical traffic signs, such as parking and way-finding signs
  • Meets ASTM Type I specifications
  • Outdoor life expectancy of 5-7 years.
3M High Intensity Prismatic sheeting
  • Highly reflective, and recommended for use in traffic, road, construction, and regulatory signs
  • Meets ASTM Type III and ASTM Type IV specifications
  • Complies with the FHWA Minimum Retroreflectivity standard
  • Outdoor life expectancy is 10 years.
The sign is compliant with MUTCD standards. All Zing signs are made in the USA from recycled materials, and support sustainability initiatives and LEED credits.
Certifications
ADA Compliance, Meets MUTCD Standards
Specifications
Specifications
White on Blue
Engineer Grade Prismatic
18in x 12in
Communication
Handicapped Parking Permit Required
English
1
Corner Holes
18in.
12in