Learn More
ZING™ Enterprises Eco Parking Sign No Parking Symbol Fire Lane Tow-Away Zone
ZING Enterprises Eco Safety sign complies with state laws.
Manufacturer:  ZING™ Enterprises 2468
Catalog No. 18-002-175
Add to cart
Description
Description
Features:
  • Alerts drivers to take precautionary measures and ensure public safety
  • Highly visible with 3M Engineer Grade Prismatic Reflective Sheeting
  • Meets MUTCD Standards
  • Made from high quality 99% recycled .080 Aluminum (80% Post Consumer) that is UL Validated for recycled content which substantiates material compliance with various green codes, standards, and procurement policies including the LEED Green Building Rating System, IgCC, GSA Advantage, ASHRAE 189.1 and others.
  • Recovered materials also exceed EPA's recommended content levels for signage and are EPA CPG Listed
  • Recycled materials support sustainability initiatives and LEED Certified Building Projects
  • Includes pre-drilled holes for easy mounting
  • Made in the USA.
Certifications
Meets MUTCD Standards
Specifications
Specifications
Red, Black, White
Engineer Grade Prismatic
18in x 12in
Communication
No Parking Symbol Fire Lane Tow-Away Zone
English
1
Corner Holes
18in.
12in