Learn More
ZING™ Enterprises Eco Parking Sign Tow Away Zone No Parking
ZING Enterprises Eco Safety Sign is pre-drilled for easy mounting. Available in two grades of reflective sheeting.
Manufacturer:  ZING™ Enterprises 2497
 View more versions of this product
Catalog No. 18-002-216
Add to cart
Description
Description
3M Engineer Grade Prismatic sheeting
  • Recommended for commercial signs and non-critical traffic signs, such as parking and way-finding signs
  • Meets ASTM Type I specifications
  • Outdoor life expectancy of 5-7 years.
3M High Intensity Prismatic sheeting
  • Highly reflective, and recommended for use in traffic, road, construction, and regulatory signs
  • Meets ASTM Type III and ASTM Type IV specifications
  • Complies with the FHWA Minimum Retroreflectivity standard
  • Outdoor life expectancy is 10 years.
The sign is compliant with MUTCD standards. All Zing signs are made in the USA from recycled materials, and support sustainability initiatives and LEED credits.
Certifications
Meets MUTCD Standards
Specifications
Specifications
Red, Black, White
High Intensity Prismatic
18in x 12in
Communication
No Parking Symbol Tow Away Zone
English
1
Corner Holes
18in.
12in